1-(3,5-dimethyl-2-((4-methylbenzyl)oxy)phenyl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

356214-1G 356214-500MG 356214-5G 356214-25G
77470-216EA 415.09 USD
77470-216 77482-884 77493-318 77503-104
1-(3,5-dimethyl-2-((4-methylbenzyl)oxy)phenyl)butan-1-ol ≥95%
1-(3,5-dimethyl-2-((4-methylbenzyl)oxy)phenyl)butan-1-ol

Precaución: For research use only

Formula: C₂₀H₂₆O₂
MW: 298.419 g/mol
Storage Temperature: Ambient
CAS Number: 3002430-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR