1-(3,5-dimethyl-2-((4-methylbenzyl)oxy)phenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

356209-1G 356209-500MG 356209-5G 356209-25G
77470-212EA 415.09 USD
77470-212 77482-880 77493-314 77503-100
1-(3,5-dimethyl-2-((4-methylbenzyl)oxy)phenyl)-2,2-difluoroethanone ≥95%
1-(3,5-dimethyl-2-((4-methylbenzyl)oxy)phenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₈H₁₈F₂O₂
Storage Temperature: Ambient
CAS Number: 3002451-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR