(3,5-Dimethyl-2-((4-methylbenzyl)oxy)phenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

356205-1G 356205-500MG 356205-5G 356205-25G
77470-208EA 415.09 USD
77470-208 77482-876 77493-310 77503-096
(3,5-Dimethyl-2-((4-methylbenzyl)oxy)phenyl)triethylsilane ≥95%
(3,5-Dimethyl-2-((4-methylbenzyl)oxy)phenyl)triethylsilane

Precaución: For research use only

Formula: C₂₂H₃₂OSi
Storage Temperature: Ambient
CAS Number: 3002528-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR