(3,5-Dimethyl-2-((4-methylbenzyl)oxy)phenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

356200-1G 356200-500MG 356200-5G 356200-25G
77470-204EA 415.09 USD
77470-204 77482-872 77493-306 77503-092
(3,5-Dimethyl-2-((4-methylbenzyl)oxy)phenyl)trimethylsilane ≥95%
(3,5-Dimethyl-2-((4-methylbenzyl)oxy)phenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₉H₂₆OSi
Storage Temperature: Ambient
CAS Number: 3002430-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR