4-Butyl-α-ethenylbenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

355977-1G 355977-500MG 355977-5G 355977-25G
77470-196EA 382.13 USD
77470-196 77482-864 77493-298 77503-084
4-Butyl-α-ethenylbenzenemethanol ≥95%
4-Butyl-α-ethenylbenzenemethanol

Precaución: For research use only

Formula: C₁₃H₁₈O
Storage Temperature: Ambient
CAS Number: 1540365-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR