3-Iodo-2-methoxy-4-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

35562-1G 35562-500MG 35562-5G 35562-25G
77470-192EA 296.64 USD
77470-192 77482-860 77493-294 77503-080
3-Iodo-2-methoxy-4-methylpyridine ≥95%
3-Iodo-2-methoxy-4-methylpyridine

Precaución: For research use only

Formula: C₇H₈INO
MW: 249.049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611478
CAS Number: 1227515-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR