2,2-Dimethyl-1-(4-(tert-pentyl)phenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

355354-1G 355354-500MG 355354-5G 355354-25G
77470-188EA 397.58 USD
77470-188 77482-856 77493-290 77503-074
2,2-Dimethyl-1-(4-(tert-pentyl)phenyl)propan-1-ol ≥95%
2,2-Dimethyl-1-(4-(tert-pentyl)phenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₆H₂₆O
Storage Temperature: Ambient
CAS Number: 1507901-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR