1-(4-(tert-pentyl)phenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

355347-1G 355347-500MG 355347-5G 355347-25G
77470-184EA 415.09 USD
77470-184 77482-852 77493-286 77503-070
1-(4-(tert-pentyl)phenyl)cyclohexanol ≥95%
1-(4-(tert-pentyl)phenyl)cyclohexanol

Precaución: For research use only

Formula: C₁₇H₂₆O
MW: 246.388 g/mol
Storage Temperature: Ambient
CAS Number: 3002481-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR