phenyl(4'-propyl-[1,1'-biphenyl]-4-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

355309-1G 355309-500MG 355309-5G 355309-25G
77470-180EA 415.09 USD
77470-180 77482-848 77493-282 77503-066
phenyl(4'-propyl-[1,1'-biphenyl]-4-yl)methanol ≥95%
phenyl(4'-propyl-[1,1'-biphenyl]-4-yl)methanol

Precaución: For research use only

Formula: C₂₂H₂₂O
MW: 302.409 g/mol
Storage Temperature: Ambient
CAS Number: 3002488-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR