1-(4'-propyl-[1,1'-biphenyl]-4-yl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

355303-1G 355303-500MG 355303-5G 355303-25G
77470-176EA 415.09 USD
77470-176 77482-842 77493-276 77503-062
1-(4'-propyl-[1,1'-biphenyl]-4-yl)propan-1-ol ≥95%
1-(4'-propyl-[1,1'-biphenyl]-4-yl)propan-1-ol

Precaución: For research use only

Formula: C₁₈H₂₂O
MW: 254.367 g/mol
Storage Temperature: Ambient
CAS Number: 3002522-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR