(5-Chloro-2-fluoro-3-methylphenyl)(m-tolyl)methanol ≥95%

Supplier: AOBChem USA

Warning

355300-1G 355300-500MG 355300-5G 355300-25G
77470-172EA 415.09 USD
77470-172 77482-840 77493-274 77503-060
(5-Chloro-2-fluoro-3-methylphenyl)(m-tolyl)methanol ≥95%
(5-Chloro-2-fluoro-3-methylphenyl)(m-tolyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₄ClFO
MW: 264.722 g/mol
Storage Temperature: Ambient
CAS Number: 3002450-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR