1-(5-Chloro-2-fluoro-3-methylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

355292-1G 355292-500MG 355292-5G 355292-25G
77470-160EA 415.09 USD
77470-160 77482-828 77493-262 77503-048
1-(5-Chloro-2-fluoro-3-methylphenyl)propan-1-one ≥95%
1-(5-Chloro-2-fluoro-3-methylphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₀H₁₀ClFO
MW: 200.637 g/mol
Storage Temperature: Ambient
CAS Number: 3002522-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR