[1,1'-biphenyl]-4-yl(3-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

355272-1G 355272-500MG 355272-5G 355272-25G
77470-156EA 415.09 USD
77470-156 77482-824 77493-258 77503-044
[1,1'-biphenyl]-4-yl(3-methoxyphenyl)methanol ≥95%
[1,1'-biphenyl]-4-yl(3-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₂₀H₁₈O₂
MW: 290.3557 g/mol
Storage Temperature: Ambient
CAS Number: 147678-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR