4-Cyano-3,5-dimethylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

35466-1G 35466-500MG 35466-5G 35466-25G
77470-152EA 287.37 USD
77470-152 77482-820 77493-252 77503-040
4-Cyano-3,5-dimethylphenylboronic acid pinacol ester ≥97%
4-Cyano-3,5-dimethylphenylboronic acid pinacol ester

Precaución: For research use only

Formula: C₁₅H₂₀BNO₂
MW: 257.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405371
CAS Number: 1111096-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR