6-Iodo-2-methylquinoline ≥95%

Supplier: AOBChem USA

Warning

354488-1G 354488-500MG 354488-5G
77470-148EA 678.77 USD
77470-148 77482-816 77493-248
6-Iodo-2-methylquinoline ≥95%
6-Iodo-2-methylquinoline

Precaución: For research use only

Formula: C₁₀H₈IN
MW: 269.08 g/mol
Storage Temperature: Ambient
CAS Number: 1128-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR