2-Chloro-4-(dimethylcarbamoyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

35410-1G 35410-500MG 35410-5G
77470-144EA 704.52 USD
77470-144 77482-812 77493-244
2-Chloro-4-(dimethylcarbamoyl)phenylboronic acid ≥95%
2-Chloro-4-(dimethylcarbamoyl)phenylboronic acid

Precaución: For research use only

Formula: C₉H₁₁BClNO₃
MW: 227.453 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22125070
CAS Number: 1449133-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR