2,4-Dichloro-5-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

35355-1G 35355-500MG 35355-5G
77470-140EA 512.94 USD
77470-140 77482-808 77493-240
2,4-Dichloro-5-methylbenzyl alcohol ≥97%
2,4-Dichloro-5-methylbenzyl alcohol

Precaución: For research use only

Formula: C₈H₈Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD28786860
CAS Number: 90003-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR