2-(4'-methyl-[1,1'-biphenyl]-4-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

353393-1G 353393-500MG 353393-5G 353393-25G
77470-136EA 415.09 USD
77470-136 77482-804 77493-236 77503-034
2-(4'-methyl-[1,1'-biphenyl]-4-yl)propan-2-ol ≥95%
2-(4'-methyl-[1,1'-biphenyl]-4-yl)propan-2-ol

Precaución: For research use only

Formula: C₁₆H₁₈O
MW: 226.314 g/mol
Storage Temperature: Ambient
CAS Number: 94571-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR