2-Methyl-1-(4'-methyl-[1,1'-biphenyl]-4-yl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

353377-1G 353377-500MG 353377-5G 353377-25G
77470-132EA 415.09 USD
77470-132 77482-800 77493-232 77503-030
2-Methyl-1-(4'-methyl-[1,1'-biphenyl]-4-yl)propan-1-ol ≥95%
2-Methyl-1-(4'-methyl-[1,1'-biphenyl]-4-yl)propan-1-ol

Precaución: For research use only

Formula: C₁₇H₂₀O
MW: 240.34 g/mol
Storage Temperature: Ambient
CAS Number: 3002444-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR