3-([1,1'-biphenyl]-2-yl)pentan-3-ol ≥95%

Supplier: AOBChem USA

Warning

353299-1G 353299-500MG 353299-5G 353299-25G
77470-128EA 415.09 USD
77470-128 77482-796 77493-228 77503-026
3-([1,1'-biphenyl]-2-yl)pentan-3-ol ≥95%
3-([1,1'-biphenyl]-2-yl)pentan-3-ol

Precaución: For research use only

Formula: C₁₇H₂₀O
MW: 240.34 g/mol
Storage Temperature: Ambient
CAS Number: 855253-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR