4-([1,1'-biphenyl]-3-yl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

353289-1G 353289-500MG 353289-5G 353289-25G
77470-124EA 415.09 USD
77470-124 77482-792 77493-224 77503-022
4-([1,1'-biphenyl]-3-yl)tetrahydro-2H-pyran-4-ol ≥95%
4-([1,1'-biphenyl]-3-yl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₇H₁₈O₂
MW: 254.324 g/mol
Storage Temperature: Ambient
CAS Number: 3002506-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR