[1,1'-biphenyl]-3-yl(4-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

353286-1G 353286-500MG 353286-5G 353286-25G
77470-120EA 415.09 USD
77470-120 77482-788 77493-220 77503-018
[1,1'-biphenyl]-3-yl(4-fluorophenyl)methanol ≥95%
[1,1'-biphenyl]-3-yl(4-fluorophenyl)methanol

Precaución: For research use only

Formula: C₁₉H₁₅FO
MW: 278.32 g/mol
Storage Temperature: Ambient
CAS Number: 3002487-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR