1-([1,1'-biphenyl]-3-yl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

353265-1G 353265-500MG 353265-5G 353265-25G
77470-116EA 415.09 USD
77470-116 77482-784 77493-216 77503-014
1-([1,1'-biphenyl]-3-yl)butan-1-ol ≥95%
1-([1,1'-biphenyl]-3-yl)butan-1-ol

Precaución: For research use only

Formula: C₁₆H₁₈O
MW: 226.314 g/mol
Storage Temperature: Ambient
CAS Number: 3002459-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR