1-([1,1'-biphenyl]-3-yl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

353259-1G 353259-500MG 353259-5G 353259-25G
77470-112EA 397.58 USD
77470-112 77482-780 77493-212 77503-010
1-([1,1'-biphenyl]-3-yl)propan-1-ol ≥95%
1-([1,1'-biphenyl]-3-yl)propan-1-ol

Precaución: For research use only

Formula: C₁₅H₁₆O
MW: 212.287 g/mol
Storage Temperature: Ambient
CAS Number: 2341905-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR