1-(2-Bromo-3-fluoropyridin-4-yl)-2-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

353123-1G 353123-500MG 353123-5G 353123-25G
77470-108EA 415.09 USD
77470-108 77482-776 77493-208 77503-006
1-(2-Bromo-3-fluoropyridin-4-yl)-2-methylbutan-1-ol ≥95%
1-(2-Bromo-3-fluoropyridin-4-yl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₃BrFNO
Storage Temperature: Ambient
CAS Number: 3002507-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR