4-Bromo-3-chloro-2-fluorothioanisole ≥95%

Supplier: AOBChem USA

Warning

35284-1G 35284-500MG 35284-5G 35284-25G
77470-104EA 109.18 USD
77470-104 77482-772 77493-204 77503-002
4-Bromo-3-chloro-2-fluorothioanisole ≥95%
4-Bromo-3-chloro-2-fluorothioanisole

Precaución: For research use only

Formula: C₇H₅BrClFS
Storage Temperature: Ambient
MDL Number: MFCD31805998
CAS Number: 214057-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR