(6-Bromo-2-fluoro-3-methylphenyl)(4-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

352019-1G 352019-500MG 352019-5G 352019-25G
77470-100EA 415.09 USD
77470-100 77482-768 77493-200 77502-998
(6-Bromo-2-fluoro-3-methylphenyl)(4-methoxyphenyl)methanol ≥95%
(6-Bromo-2-fluoro-3-methylphenyl)(4-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₄BrFO₂
MW: 325.1728 g/mol
Storage Temperature: Ambient
CAS Number: 3002507-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR