(6-Bromo-2-fluoro-3-methylphenyl)(m-tolyl)methanol ≥95%

Supplier: AOBChem USA

Warning

352016-1G 352016-500MG 352016-5G 352016-25G
77470-096EA 415.09 USD
77470-096 77482-764 77493-196 77502-994
(6-Bromo-2-fluoro-3-methylphenyl)(m-tolyl)methanol ≥95%
(6-Bromo-2-fluoro-3-methylphenyl)(m-tolyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₄BrFO
MW: 309.1734 g/mol
Storage Temperature: Ambient
CAS Number: 3002520-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR