4-(Dimethylamino)-2-(trifluoromethyl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

351787-1G 351787-500MG 351787-5G 351787-25G
77470-092EA 126.69 USD
77470-092 77482-760 77493-192 77502-990
4-(Dimethylamino)-2-(trifluoromethyl)benzonitrile ≥95%
4-(Dimethylamino)-2-(trifluoromethyl)benzonitrile

Precaución: For research use only

Formula: C₁₀H₉F₃N₂
MW: 214.19 g/mol
Storage Temperature: Ambient
CAS Number: 51332-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR