3-Bromo-5-fluoro-2-methylphenol ≥95%

Supplier: AOBChem USA

Warning

35041-1G 35041-500MG 35041-5G
77470-088EA 1050.6 USD
77470-088 77482-756 77493-186
3-Bromo-5-fluoro-2-methylphenol ≥95%
3-Bromo-5-fluoro-2-methylphenol

Precaución: For research use only

Formula: C₇H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD28527221
CAS Number: 1784522-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR