2-Bromo-6-chloro-3-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

34960-1G 34960-500MG 34960-5G 34960-25G
77470-084EA 284.28 USD
77470-084 77482-752 77493-180 77502-980
2-Bromo-6-chloro-3-fluorophenol ≥97%
2-Bromo-6-chloro-3-fluorophenol

Precaución: For research use only

Formula: C₆H₃BrClFO
MW: 225.443 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414731
CAS Number: 1782417-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR