4-Bromo-2-chloro-3-ethoxytoluene ≥95%

Supplier: AOBChem USA

Warning

34919-1G 34919-500MG 34919-5G 34919-25G
77470-080EA 151.41 USD
77470-080 77482-748 77493-176 77502-976
4-Bromo-2-chloro-3-ethoxytoluene ≥95%
4-Bromo-2-chloro-3-ethoxytoluene

Precaución: For research use only

Formula: C₉H₁₀BrClO
Storage Temperature: Ambient
MDL Number: MFCD31805779
CAS Number: 2244107-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR