2-Chloro-2,2-difluoro-1-(4'-propyl[1,1'-biphenyl]-4-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

348485-1G 348485-500MG 348485-5G
77470-076EA 576.8 USD
77470-076 77482-744 77493-172
2-Chloro-2,2-difluoro-1-(4'-propyl[1,1'-biphenyl]-4-yl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(4'-propyl[1,1'-biphenyl]-4-yl)ethanone

Precaución: For research use only

Formula: C₁₇H₁₅ClF₂O
MW: 308.75 g/mol
Storage Temperature: Ambient
CAS Number: 2173094-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR