Cyclohexyl(4'-propyl[1,1'-biphenyl]-4-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

348476-1G 348476-500MG 348476-5G
77470-072EA 576.8 USD
77470-072 77482-740 77493-168
Cyclohexyl(4'-propyl[1,1'-biphenyl]-4-yl)methanone ≥95%
Cyclohexyl(4'-propyl[1,1'-biphenyl]-4-yl)methanone

Precaución: For research use only

Formula: C₂₂H₂₆O
Storage Temperature: Ambient
CAS Number: 2173122-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR