(4'-Methyl[1,1'-biphenyl]-4-yl)phenylmethanone ≥95%

Supplier: AOBChem USA

Warning

348418-1G 348418-500MG 348418-5G 348418-25G
77470-068EA 415.09 USD
77470-068 77482-736 77493-164 77502-970
(4'-Methyl[1,1'-biphenyl]-4-yl)phenylmethanone ≥95%
(4'-Methyl[1,1'-biphenyl]-4-yl)phenylmethanone

Precaución: For research use only

Formula: C₂₀H₁₆O
MW: 272.34 g/mol
Storage Temperature: Ambient
CAS Number: 63283-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR