4-Bromo-2,6-dimethylbiphenyl ≥97%

Supplier: AOBChem USA

Warning

34799-1G 34799-500MG 34799-5G 34799-25G
77470-064EA 255.44 USD
77470-064 77482-732 77493-160 77502-966
4-Bromo-2,6-dimethylbiphenyl ≥97%
4-Bromo-2,6-dimethylbiphenyl

Precaución: For research use only

Formula: C₁₄H₁₃Br
Storage Temperature: Ambient
MDL Number: MFCD31806004
CAS Number: 958650-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR