5-Fluoro-2-isopropoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

34745-1G 34745-500MG 34745-5G 34745-25G
77470-060EA 226.6 USD
77470-060 77482-728 77493-156 77502-964
5-Fluoro-2-isopropoxybenzaldehyde ≥95%
5-Fluoro-2-isopropoxybenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11639864
CAS Number: 610797-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR