Methyl 4-bromothiazole-5-carboxylate ≥97%

Supplier: AOBChem USA

Warning

34701-1G 34701-500MG 34701-5G 34701-25G
77470-056EA 105.06 USD
77470-056 77482-726 77493-154 77502-960
Methyl 4-bromothiazole-5-carboxylate ≥97%
Methyl 4-bromothiazole-5-carboxylate

Precaución: For research use only

Formula: C₅H₄BrNO₂S
Storage Temperature: Ambient
MDL Number: MFCD11111597
CAS Number: 1353100-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR