(4-Isobutoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

347010-1G 347010-500MG 347010-5G 347010-25G
77470-052EA 415.09 USD
77470-052 77482-720 77493-148 77502-956
(4-Isobutoxyphenyl)(phenyl)methanol ≥95%
(4-Isobutoxyphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₇H₂₀O₂
MW: 256.3395 g/mol
Storage Temperature: Ambient
CAS Number: 93727-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR