2,2-Difluoro-1-(4-isobutoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

347008-1G 347008-500MG 347008-5G 347008-25G
77470-048EA 415.09 USD
77470-048 77482-716 77493-144 77502-952
2,2-Difluoro-1-(4-isobutoxyphenyl)ethanone ≥95%
2,2-Difluoro-1-(4-isobutoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₄F₂O₂
MW: 228.2351 g/mol
Storage Temperature: Ambient
CAS Number: 3002454-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR