2-(3-((2-fluorobenzyl)oxy)-5-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

346829-1G 346829-500MG 346829-5G 346829-25G
77470-044EA 415.09 USD
77470-044 77482-712 77493-140 77502-948
2-(3-((2-fluorobenzyl)oxy)-5-methylphenyl)propan-2-ol ≥95%
2-(3-((2-fluorobenzyl)oxy)-5-methylphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₇H₁₉FO₂
MW: 274.33 g/mol
Storage Temperature: Ambient
CAS Number: 3002535-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR