(6-Bromo-2-fluoro-3-methylphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

346700-1G 346700-500MG 346700-5G 346700-25G
77470-036EA 415.09 USD
77470-036 77482-704 77493-130 77502-938
(6-Bromo-2-fluoro-3-methylphenyl)triethylsilane ≥95%
(6-Bromo-2-fluoro-3-methylphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₃H₂₀BrFSi
MW: 303.286 g/mol
Storage Temperature: Ambient
CAS Number: 3002523-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR