(6-Bromo-2-fluoro-3-methylphenyl)(thiophen-2-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

346694-1G 346694-500MG 346694-5G 346694-25G
77470-032EA 415.09 USD
77470-032 77482-700 77493-126 77502-934
(6-Bromo-2-fluoro-3-methylphenyl)(thiophen-2-yl)methanol ≥95%
(6-Bromo-2-fluoro-3-methylphenyl)(thiophen-2-yl)methanol

Precaución: For research use only

Formula: C₁₂H₁₀BrFOS
MW: 301.175 g/mol
Storage Temperature: Ambient
CAS Number: 3002476-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR