(6-Bromo-2-fluoro-3-methylphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

346680-1G 346680-500MG 346680-5G 346680-25G
77470-028EA 415.09 USD
77470-028 77482-696 77493-122 77502-930
(6-Bromo-2-fluoro-3-methylphenyl)(cyclopropyl)methanol ≥95%
(6-Bromo-2-fluoro-3-methylphenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₁H₁₂BrFO
Storage Temperature: Ambient
CAS Number: 3002460-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR