1-(3-Bromobenzo[b]thiophen-2-yl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

Warning

346676-1G 346676-500MG 346676-5G 346676-25G
77470-024EA 415.09 USD
77470-024 77482-692 77493-118 77502-926
1-(3-Bromobenzo[b]thiophen-2-yl)-2-methylpropan-1-one ≥95%
1-(3-Bromobenzo[b]thiophen-2-yl)-2-methylpropan-1-one

Precaución: For research use only

Formula: C₁₂H₁₁BrOS
MW: 283.184 g/mol
Storage Temperature: Ambient
CAS Number: 3002443-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR