1-(3-Bromobenzo[b]thiophen-2-yl)-1-phenylethanol ≥95%

Supplier: AOBChem USA

Warning

346674-1G 346674-500MG 346674-5G 346674-25G
77470-020EA 415.09 USD
77470-020 77482-688 77493-114 77502-922
1-(3-Bromobenzo[b]thiophen-2-yl)-1-phenylethanol ≥95%
1-(3-Bromobenzo[b]thiophen-2-yl)-1-phenylethanol

Precaución: For research use only

Formula: C₁₆H₁₃BrOS
Storage Temperature: Ambient
CAS Number: 3002523-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR