(3-Bromobenzo[b]thiophen-2-yl)(2,4-dichlorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

346672-1G 346672-500MG 346672-5G 346672-25G
77470-016EA 415.09 USD
77470-016 77482-684 77493-110 77502-918
(3-Bromobenzo[b]thiophen-2-yl)(2,4-dichlorophenyl)methanol ≥95%
(3-Bromobenzo[b]thiophen-2-yl)(2,4-dichlorophenyl)methanol

Precaución: For research use only

Formula: C₁₅H₉BrCl₂OS
Storage Temperature: Ambient
CAS Number: 3002487-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR