1-(3-Bromobenzo[b]thiophen-2-yl)-1-cyclopropylethanol ≥95%

Supplier: AOBChem USA

Warning

346670-1G 346670-500MG 346670-5G 346670-25G
77470-012EA 415.09 USD
77470-012 77482-680 77493-106 77502-914
1-(3-Bromobenzo[b]thiophen-2-yl)-1-cyclopropylethanol ≥95%
1-(3-Bromobenzo[b]thiophen-2-yl)-1-cyclopropylethanol

Precaución: For research use only

Formula: C₁₃H₁₃BrOS
Storage Temperature: Ambient
CAS Number: 3002506-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR