2-(3-Bromobenzo[b]thiophen-2-yl)butan-2-ol ≥95%

Supplier: AOBChem USA

Warning

346668-1G 346668-500MG 346668-5G 346668-25G
77470-008EA 415.09 USD
77470-008 77482-676 77493-102 77502-910
2-(3-Bromobenzo[b]thiophen-2-yl)butan-2-ol ≥95%
2-(3-Bromobenzo[b]thiophen-2-yl)butan-2-ol

Precaución: For research use only

Formula: C₁₂H₁₃BrOS
MW: 285.2 g/mol
Storage Temperature: Ambient
CAS Number: 3002453-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR