(3-Bromobenzo[b]thiophen-2-yl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

346665-1G 346665-500MG 346665-5G 346665-25G
77470-004EA 415.09 USD
77470-004 77482-672 77493-098 77502-906
(3-Bromobenzo[b]thiophen-2-yl)(cyclopropyl)methanol ≥95%
(3-Bromobenzo[b]thiophen-2-yl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₂H₁₁BrOS
Storage Temperature: Ambient
CAS Number: 3002439-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR